2,4-Dibromopyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR8632-25G OR8632-5G
77411-416EA 41.78 USD
77411-416 77411-418
2,4-Dibromopyridine 97%
2,4-Dibromopyridine
Formula: C₅H₃Br₂N
Melting Pt: 35…40 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01859720
CAS Number: 58530-53-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR