3-Bromothiophene 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR6570-100G OR6570-25G OR6570-500G
77410-864EA 69.28 USD
77410-864 77410-866 77410-868
3-Bromothiophene 97%
3-Bromothiophene
Formula: C₄H₃BrS
Boiling Pt: 92…93 °C/90 mmHg
Density: 1.722 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005464
CAS Number: 872-31-1
EINECS: 212-821-3
UN: 2929
ADR: 6.1+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR