2-Bromopyridine

Supplier: APOLLO SCIENTIFIC
Danger

OR23057-100G OR23057-1KG OR23057-50G
77407-050EA 34.64 USD
77407-050 77407-052 77407-054
2-Bromopyridine
2-Bromopyridine
Formula: C₅H₄BrN
MW: 158 g/mol
Boiling Pt: 192
Melting Pt: -40.1
Density: 1.657 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00006219
CAS Number: 109-04-6
UN: 2810
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR