2,6-Dibromopyridine

Supplier: APOLLO SCIENTIFIC
Danger

OR28767-25G OR28767-100G
77408-214EA 21.26 USD
77408-214 77408-212
2,6-Dibromopyridine
2,6-Dibromopyridine
Formula: C₅H₃Br₂N
Boiling Pt: 255 °C
Melting Pt: 118…119 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006223
CAS Number: 626-05-1
EINECS: 210-926-9
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR