Silver fluoride 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Silver monofluoride, Silver(I) fluoride, Argentous fluoride

PC6370-25G PC6370-5G
77417-362EA 296.48 USD
77417-362 77417-364
Silver fluoride 98%
Silver fluoride

Soft (mild) fluorinating agent for selective fluorination. Alkyl fluorides may be prepared by halex reactions. AgF & I2 or Br2 will trans-fluoro halogenate alkenes.

Formula: AgF
MW: 126.87 g/mol
Boiling Pt: 1159 °C
Melting Pt: >300 °C
Density: 5.852 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003410
CAS Number: 7775-41-9
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR