4-Bromoisoquinoline

Supplier: APOLLO SCIENTIFIC
Warning

OR14319-25G OR14319-5G
77404-904EA 32.6 USD
77404-904 77404-906
4-Bromoisoquinoline
4-Bromoisoquinoline
Formula: C₉H₆BrN
Boiling Pt: 280…285 °C
Melting Pt: 40…43 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006904
CAS Number: 1532-97-4
EINECS: 216-244-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR