Sodium tetrafluoroborate 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC6560-100G PC6560-25G PC6560-500G
77417-430EA 85.58 USD
77417-430 77417-432 77417-434
Sodium tetrafluoroborate 97%
Sodium tetrafluoroborate
Formula: BF₄Na
Melting Pt: 384 °C (dec.)
Density: 2.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003515
CAS Number: 13755-29-8
EINECS: 237-340-6
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR