1,2-Dichlorooctafluorocyclohexene 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC2500-1G PC2500-5G
77413-668EA 21.32 USD
77413-668 77413-670
1,2-Dichlorooctafluorocyclohexene 97%
1,2-Dichlorooctafluorocyclohexene
Formula: C₆Cl₂F₈
Boiling Pt: 112…114 °C
Density: 1.719 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039466
CAS Number: 336-19-6
EINECS: 206-408-7
UN: 2810
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR