Cycloheptanol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Hydroxycycloheptane, Suberyl alcohol

OR10091-25G OR10091-5G
77402-466EA 63.16 USD
77402-466 77402-468
Cycloheptanol
Cycloheptanol
Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 185 °C
Density: 0.948 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00004150
CAS Number: 502-41-0
EINECS: 207-936-0
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR