1,2-Cyclohexanedione 95%

Supplier: APOLLO SCIENTIFIC

OR0317-25G OR0317-5G
77402-016EA 146.7 USD
77402-016 77402-018
1,2-Cyclohexanedione 95%
1,2-Cyclohexanedione
Formula: C₆H₈O₂
Boiling Pt: 193…195 °C
Melting Pt: 34…38 °C
Density: 1.12 g/cm³ (25 °C)
MDL Number: MFCD00001648
CAS Number: 765-87-7
EINECS: 212-155-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR