2-Methyl-8-quinolinol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 8-Hydroxy-2-methylquinoline, 8-Hydroxyquinaldine, 2-Methyloxine

OR2004-100G OR2004-25G
77406-542EA 52.98 USD
77406-542 77406-544
2-Methyl-8-quinolinol 98%
2-Methyl-8-quinolinol
Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 267 °C
Melting Pt: 71…73 °C
Flash Pt: 139 °C
Storage Temperature: Ambient
MDL Number: MFCD00006765
CAS Number: 826-81-3
EINECS: 212-562-6
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR