Cloxiquina 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR1049-100G OR1049-500G
77402-742EA 23.44 USD
77402-742 77402-752
Cloxiquina 98%
Cloxiquina
Formula: C₉H₆ClNO
Melting Pt: 121…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00006788
CAS Number: 130-16-5
EINECS: 204-978-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR