4-Quinolinecarbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Quinoline-4-carbaldehyde, Quinoline-4-carboxaldehyde, 4-Quinolinecarboxaldehyde

OR23077-25G OR23077-5G OR23077-1G
77407-074EA 174.22 USD
77407-074 77407-076 77407-072
4-Quinolinecarbaldehyde
4-Quinolinecarbaldehyde
Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 45…52 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006781
CAS Number: 4363-93-3
EINECS: 224-453-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR