2,4,5-Trimethylaniline

Supplier: APOLLO SCIENTIFIC
Danger

OR27602-250MG OR27602-5G OR27602-1G
77407-954EA 33.62 USD
77407-954 77407-956 77407-952
2,4,5-Trimethylaniline
2,4,5-Trimethylaniline
Formula: C₉H₁₃N
MW: 135.21 g/mol
Melting Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00025201
CAS Number: 137-17-7
EINECS: 205-282-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR