2,6-Dimethylthiophenol 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR451316-100G OR451316-25G OR451316-5G
77409-796EA 343.2 USD
77409-796 77409-798 77409-800
2,6-Dimethylthiophenol 95+%
2,6-Dimethylthiophenol
Formula: C₈H₁₀S
Boiling Pt: 122 °C
Density: 1.038 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00010021
CAS Number: 118-72-9
UN: 3334
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR