α,α,α-Trifluoro-p-xylene 96+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Trifluoromethyl)toulene, 1-Methyl-4-(trifluoromethyl)benzene, 4-Methylbenzotrifluoride

PC5067C-100G PC5067C-25G PC5067C-5G
77416-394EA 399.38 USD
77416-394 77416-396 77416-398
α,α,α-Trifluoro-p-xylene 96+%
α,α,α-Trifluoro-p-xylene
Formula: C₈H₇F₃
MW: 160.14 g/mol
Boiling Pt: 129 °C
Density: 1.144 g/cm³ (25 °C)
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00075476
CAS Number: 6140-17-6
EINECS: 228-124-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR