3-Fluoro-o-xylene 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC4381C-100G
PC4381C-250G
PC4381C-25G
77415-384EA
124.1
USD
77415-384
77415-386
77415-388
3-Fluoro-o-xylene 98%
3-Fluoro-o-xylene
Formula:
C₈H₉F Boiling Pt: 147…152 °C Density: 0.992 g/cm³ (25 °C) Flash Pt: 36 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000323 CAS Number: 443-82-3 EINECS: 207-140-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...