α'-Bromo-α,α,α-trifluoro-p-xylene 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC7633-25G PC7633-5G
77418-050EA 23.28 USD
77418-050 77418-052
α'-Bromo-α,α,α-trifluoro-p-xylene 98%
α'-Bromo-α,α,α-trifluoro-p-xylene
Formula: C₈H₆BrF₃
Boiling Pt: 65…66 °C/5 mmHg
Melting Pt: 30…31 °C
Density: 1.546 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00000403
CAS Number: 402-49-3
EINECS: 206-947-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR