4-Nitro-α,α,α-trifluorotoluene 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC5252-10G
PC5252-1G
PC5252-5G
77416-566EA
96.78
USD
77416-566
77416-568
77416-570
4-Nitro-α,α,α-trifluorotoluene 98%
4-Nitro-α,α,α-trifluorotoluene
Formula:
C₇H₄F₃NO₂ Boiling Pt: 81…83 °C/10 mmHg Melting Pt: 38…40 °C Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007358 CAS Number: 402-54-0 EINECS: 206-948-3 UN: 3431 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...