2-Nitro-α,α,α-trifluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC5240-100G PC5240-25G
77416-540EA 247.58 USD
77416-540 77416-542
2-Nitro-α,α,α-trifluorotoluene 98%
2-Nitro-α,α,α-trifluorotoluene
Formula: C₇H₄F₃NO₂
Boiling Pt: 104…105 °C/20 mmHg
Melting Pt: 31…32 °C
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00007145
CAS Number: 384-22-5
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR