4-Fluoro-3-nitrobenzotrifluoride 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Fluoro-3-Nitrobenzotrifluoride

PC3950-100G PC3950-25G PC3950-500G
77415-022EA 89.66 USD
77415-022 77415-024 77415-026
4-Fluoro-3-nitrobenzotrifluoride 98%
4-Fluoro-3-nitrobenzotrifluoride
Formula: C₇H₃F₄NO₂
MW: 209.1 g/mol
Boiling Pt: 91…92 °C/15 mmHg
Density: 1.494 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00007059
CAS Number: 367-86-2
EINECS: 206-702-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR