α,α,α-Trifluoro-3-iodotoluene 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4891-100G PC4891-1KG PC4891-500G
77415-824EA 117.16 USD
77415-824 77415-826 77415-828
α,α,α-Trifluoro-3-iodotoluene 97%
α,α,α-Trifluoro-3-iodotoluene
Formula: C₇H₄F₃I
Boiling Pt: 82 °C/25 mmHg
Melting Pt: –8 °C
Density: 1.887 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00001049
CAS Number: 401-81-0
EINECS: 206-934-7
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR