2-Nitrobenzyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Chloromethyl)-2-nitrobenzene, 2-Nitrobenzyl chloride, α-Chloro-o-nitrotoluene, alpha-Chloro-2-nitrotoluene

OR46019-1G OR46019-5G
77409-874EA 21.26 USD
77409-874 77409-876
2-Nitrobenzyl chloride 98%
2-Nitrobenzyl chloride
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 127…133 °C/10 mmHg
Melting Pt: 46…48 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007185
CAS Number: 612-23-7
EINECS: 210-300-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR