3,4-Dichloro-α,α,α-trifluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,2-Dichloro-4-(trifluoromethyl)benzene, 3,4-Dichlorobenzotrifluoride

PC2340-100G PC2340-500G
77413-602EA 21.26 USD
77413-602 77413-604
3,4-Dichloro-α,α,α-trifluorotoluene 98%
3,4-Dichloro-α,α,α-trifluorotoluene
Formula: C₇H₃Cl₂F₃
MW: 215 g/mol
Boiling Pt: 173…174 °C
Melting Pt: –12 °C
Density: 1.479 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000555
CAS Number: 328-84-7
EINECS: 206-337-1
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR