(1-Chloroethyl)benzene 97%

Supplier: APOLLO SCIENTIFIC

OR9525-25ML OR9525-500ML
77412-168EA 117.16 USD
77412-168 77412-170
(1-Chloroethyl)benzene 97%
(1-Chloroethyl)benzene
Formula: C₆H₅CHClCH₃
MDL Number: MFCD00018870
CAS Number: 672-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR