4-Bromo-α,α,α-trifluorotoluene 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC1350-100G PC1350-1KG PC1350-500G
77413-134EA 54 USD
77413-134 77413-136 77413-138
4-Bromo-α,α,α-trifluorotoluene 99%
4-Bromo-α,α,α-trifluorotoluene
Formula: C₇H₄BrF₃
Boiling Pt: 154…156 °C
Density: 1.607 g/cm³ (25 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00000398
CAS Number: 402-43-7
EINECS: 206-943-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR