α,4-Dibromo-2-fluorotoluene 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Bromo-1-(bromomethyl)-2-fluorobenzene, 4-Bromo-2-fluorobenzyl bromide

PC1423H-100G PC1423H-25G PC1423H-5G
77413-164EA 110.04 USD
77413-164 77413-166 77413-168
α,4-Dibromo-2-fluorotoluene 97%
α,4-Dibromo-2-fluorotoluene
Formula: C₇H₅Br₂F
Boiling Pt: 126 °C/19 mmHg
Melting Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00055467
CAS Number: 76283-09-5
EINECS: 278-412-7
UN: 2923
ADR: 8+6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR