α-Bromo-2-fluorotoulene 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Fluorobenzyl bromide, α-Bromo-o-fluorotoulene

PC3693-100G PC3693-250G PC3693-25G
77414-814EA 41.78 USD
77414-814 77414-816 77414-818
α-Bromo-2-fluorotoulene 97%
α-Bromo-2-fluorotoulene
Formula: C₇H₆BrF
Boiling Pt: 62…64 °C/15 mmHg
Density: 1.563 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000324
CAS Number: 446-48-0
EINECS: 207-169-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR