α-Bromo-2,5-difluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(Bromomethyl)-1,4-difluorobenzene, 2,5-Difluorobenzyl bromide

PC2742-10G PC2742-250G PC2742-50G
77413-818EA 21.26 USD
77413-818 77413-820 77413-822
α-Bromo-2,5-difluorotoluene 98%
α-Bromo-2,5-difluorotoluene
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Density: 1.609 g/cm³ (25 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00009897
CAS Number: 85117-99-3
UN: 2924
ADR: 3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR