Pentafluorobenzyl bromide 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α-Bromo-2,3,4,5,6-pentafluorotoluene, 2,3,4,5,6-Pentafluorobenzyl bromide

PC5550-100G PC5550-10G PC5550-25G
77416-786EA 470.7 USD
77416-786 77416-788 77416-790
Pentafluorobenzyl bromide 98%
Pentafluorobenzyl bromide
Formula: (C₆F₅)CH₂Br
MW: 260.99 g/mol
Boiling Pt: 174
Melting Pt: 19…20 °C
Density: 1.728 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00000299
CAS Number: 1765-40-8
EINECS: 217-182-4
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR