Benzyl bromide 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR12905-100G OR12905-25G
77404-464EA 41.78 USD
77404-464 77404-466
Benzyl bromide 99%
Benzyl bromide
Formula: (C₆H₅)CH₂Br
Boiling Pt: 198…199 °C
Melting Pt: –3…–1 °C
Density: 1.438 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00000172
CAS Number: 100-39-0
EINECS: 202-847-3
UN: 1737
ADR: 6.1+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR