N-Cyclohexylaniline

Supplier: APOLLO SCIENTIFIC
Warning

OR28787-10G OR28787-1G OR28787-5G
77408-242EA 96.78 USD
77408-242 77408-244 77408-246
N-Cyclohexylaniline
N-Cyclohexylaniline
Formula: C₁₂H₁₇N
Boiling Pt: 279 °C
Melting Pt: 14…15 °C
Density: 0.996 g/cm³ (25 °C)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00014284
CAS Number: 1821-36-9
EINECS: 217-344-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR