N,N-Diethyl-4-nitroaniline 98%

Supplier: APOLLO SCIENTIFIC

OR962461-1G OR962461-25G OR962461-5G
77412-242EA 33.62 USD
77412-242 77412-244 77412-246
N,N-Diethyl-4-nitroaniline 98%
N,N-Diethyl-4-nitroaniline
Formula: C₁₀H₁₄N₂O₂
Melting Pt: 77.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00043606
CAS Number: 2216-15-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR