4-Fluorothiophenol 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
4-Fluorobenzene-1-thiol, 1-Fluoro-4-mercaptobenzene, 4-Fluorobenzenethiol, p-Fluorothiophenol
PC4300-100G
PC4300-250G
PC4300-25G
77415-244EA
41.78
USD
77415-244
77415-246
77415-248
4-Fluorothiophenol 98%
4-Fluorothiophenol
Intermediate for the synthesis of Bicalutamide.
Formula:
C₆H₅FS MW: 128.17 g/mol Boiling Pt: 168…172 °C Density: 1.203 g/cm³ (25 °C) Flash Pt: 54 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004846 CAS Number: 371-42-6 EINECS: 206-737-6 UN: 3071 ADR: 6.1+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...