2,5-Difluorophenol 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC2867-25G PC2867-100G
77413-934EA 75.4 USD
77413-934 77413-932
2,5-Difluorophenol 97%
2,5-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 145 °C
Melting Pt: 36…42 °C
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00042501
CAS Number: 2713-31-7
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR