4-(Trifluoromethoxy)anisole 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7434C-100G PC7434C-25G PC7434C-5G
77417-872EA 99.74 USD
77417-872 77417-874 77417-876
4-(Trifluoromethoxy)anisole 98%
4-(Trifluoromethoxy)anisole
Formula: C₈H₇F₃O₂
Boiling Pt: 163…165 °C
Density: 1.272 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00216942
CAS Number: 710-18-9
EINECS: 615-239-1
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR