2,3,6-Trifluoroanisole 98%

Supplier: APOLLO SCIENTIFIC

PC0510-250MG PC0510-1G
77412-594EA 21.26 USD
77412-594 77412-592
2,3,6-Trifluoroanisole 98%
2,3,6-Trifluoroanisole
Formula: C₇H₅F₃O
Boiling Pt: 146 °C
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD01631501
CAS Number: 4920-34-7
UN: 3271
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR