2-Fluoroanisole 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC3420-100G PC3420-500G
77414-722EA 21.34 USD
77414-722 77414-724
2-Fluoroanisole 99%
2-Fluoroanisole
Formula: C₇H₇FO
Boiling Pt: 154…155 °C
Density: 1.124 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00000316
CAS Number: 321-28-8
EINECS: 206-284-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR