2,6-Difluoroanisole 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC0537-100G PC0537-10G PC0537-25G
77412-618EA 339.26 USD
77412-618 77412-620 77412-622
2,6-Difluoroanisole 98%
2,6-Difluoroanisole
Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 70…72 °C/56 mmHg
Density: 1.221 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00142846
CAS Number: 437-82-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR