1,2,4-Trifluoro-5-nitrobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,4,5-Trifluoronitrobenzene

PC7790-100G PC7790-1KG PC7790-250G
77418-202EA 63.16 USD
77418-202 77418-204 77418-206
1,2,4-Trifluoro-5-nitrobenzene 98%
1,2,4-Trifluoro-5-nitrobenzene
Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
Boiling Pt: 194…195 °C/760 mmHg
Melting Pt: –11 °C
Density: 1.544 g/cm³ (25 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00007051
CAS Number: 2105-61-5
EINECS: 218-281-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR