3-Fluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3540-100G PC3540-25G PC3540-500G
77414-758EA 49.92 USD
77414-758 77414-760 77414-762
3-Fluorobenzonitrile 98%
3-Fluorobenzonitrile
Formula: C₇H₄FN
Boiling Pt: 182…183 °C
Melting Pt: 15…16 °C
Density: 1.134 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00001797
CAS Number: 403-54-3
EINECS: 206-963-5
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR