2,5-Difluorobenzaldehyde 95%

Supplier: APOLLO SCIENTIFIC

PC48832-100G PC48832-250G PC48832-25G
77415-806EA 83.54 USD
77415-806 77415-808 77415-810
2,5-Difluorobenzaldehyde 95%
2,5-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010327
CAS Number: 2646-90-4
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR