1,2,4,5-Tetrafluorobenzene 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC6730-10G PC6730-250G PC6730-50G
77417-514EA 21.26 USD
77417-514 77417-516 77417-518
1,2,4,5-Tetrafluorobenzene 99%
1,2,4,5-Tetrafluorobenzene
Formula: C₆H₂F₄
MW: 150.08 g/mol
Boiling Pt: 90 °C
Melting Pt: 3…5 °C
Density: 1.344 g/cm³ (25 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00000307
CAS Number: 327-54-8
EINECS: 206-319-3
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR