4-Chlorothiophenol 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Chloro-4-mercaptobenzene, 4-Chlorobenzene-1-thiol, 4-Chlorobenzenethiol, p-Chlorothiophenol

OR54321-100G OR54321-25G OR54321-500G
77410-360EA 48.9 USD
77410-360 77410-362 77410-364
4-Chlorothiophenol 99%
4-Chlorothiophenol
Formula: C₆H₅ClS
MW: 144.62 g/mol
Boiling Pt: 205…207 °C
Melting Pt: 48…52 °C
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004847
CAS Number: 106-54-7
EINECS: 203-408-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR