2-Chlorothiophenol 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR1590-100G OR1590-25G OR1590-5G
77405-572EA 92 USD
77405-572 77405-574 77405-576
2-Chlorothiophenol 99%
2-Chlorothiophenol
Formula: C₆H₅ClS
Boiling Pt: 204…206 °C
Density: 1.275 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004830
CAS Number: 6320-03-2
EINECS: 228-667-5
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR