5-Chloro-2-nitroaniline

Supplier: APOLLO SCIENTIFIC
Danger

OR28701-10G OR28701-25G OR28701-100G
77408-174EA 21.26 USD
77408-174 77408-176 77408-172
5-Chloro-2-nitroaniline
5-Chloro-2-nitroaniline
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Boiling Pt: 323
Melting Pt: 124…129 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007776
CAS Number: 1635-61-6
EINECS: 216-661-5
UN: 2237
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR