5-Chloro-2-fluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC0082-100G PC0082-25G PC0082-5G
77412-382EA 66.22 USD
77412-382 77412-384 77412-386
5-Chloro-2-fluoroaniline 98%
5-Chloro-2-fluoroaniline
Formula: C₆H₅ClFN
Boiling Pt: 102…105 °C/20 mmHg
Melting Pt: 20…22 °C
Density: 1.29 g/cm³ (25 °C)
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00069416
CAS Number: 2106-05-0
EINECS: 218-284-1
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR