3-Chloro-2-fluorobenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC2309-10G PC2309-25G PC2309-5G
77413-594EA 23.44 USD
77413-594 77413-596 77413-598
3-Chloro-2-fluorobenzonitrile
3-Chloro-2-fluorobenzonitrile
Formula: C₇H₃ClFN
Melting Pt: 40…44 °C
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00042206
CAS Number: 94087-40-8
EINECS: 301-934-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR