3-Chlorobenzoic acid
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR10911-100G
OR10911-25G
OR10911-500G
77402-968EA
28.52
USD
77402-968
77402-970
77402-972
3-Chlorobenzoic acid
3-Chlorobenzoic acid
Formula:
C₇H₅ClO₂ Melting Pt: 153…157 °C Density: 1.496 g/cm³ (25 °C) Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002491 CAS Number: 535-80-8 EINECS: 208-618-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...