2-Amino-4-chlorobenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR15043-50G OR15043-1KG OR15043-250G
77405-262EA 52.98 USD
77405-262 77405-258 77405-260
2-Amino-4-chlorobenzonitrile
2-Amino-4-chlorobenzonitrile
Formula: C₇H₅ClN₂
Melting Pt: 157…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00035927
CAS Number: 38487-86-4
EINECS: 253-967-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR